missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GTP Solution (100 mM)

Catalog No. FERR0461
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0461 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR0461 Supplier Thermo Scientific™ Supplier No. R0461
Only null left

Thermo Scientific GTP (guanosine 5'-triphosphate) is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH.

Thermo Scientific GTP (guanosine 5'-triphosphate) is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. It is more than 99% pure and highly stable. GTP may be used in a wide variety of different molecular biology applications.

Features
• Greater than 99% purity confirmed by HPLC
• Functionally tested in in vitro transcription
• Stable for two years at -20°C and after multiple freeze-thaw cycles

Applications
• aRNA synthesis
• siRNA synthesis
• RNA amplification
in vitro transcription

Specifications

Concentration 100 mM
For Use With (Application) in vitro Transcription, RNA Amplification
Content And Storage GTP Solution, 100 mM (0.25 mL)

Store at –20°C.
Shipping Condition Dry Ice
Conjugate Unconjugated
Quantity 0.25 mL
Product Type CTP Solution
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.