missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneRuler 100 bp Plus DNA Ladder

Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.

$110.42 - $488.75

Specifications

Concentration 6X
Gel Type Agarose
Label or Dye Xylene Cyanol FF, Bromophenol Blue, Orange G
Ready to Load No
Size Range 100 to 3000 bp
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERSM0321
View Documents
Thermo Scientific™
SM0321
50 μg
Each for $110.42
Only null left
 
FERSM0322
View Documents
Thermo Scientific™
SM0322
5 x 50 μg
Each for $488.75
Only null left
 
Description

Description

GeneRuler 100bp Plus DNA Ladder 100 to 3000bp is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.

GeneRuler 100bp Plus DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).

The ladder is supplied with 6X DNA Loading Dye.

Highlights

  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA
Specifications

Specifications

6X
Xylene Cyanol FF, Bromophenol Blue, Orange G
100 to 3000 bp
GeneRuler
Agarose
No
Agarose Gels
DNA Ladder
Videos
SDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.