Learn More
Thermo Scientific™ GeneRuler 100 bp Plus DNA Ladder
Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels.
Supplier: Thermo Scientific™ SM0322
Description
GeneRuler 100bp Plus DNA Ladder 100 to 3000bp is recommended for sizing and approximate quantification of a wide range of double-stranded DNA on agarose or polyacrylamide gels. The ladder is a mixture of chromatography-purified individual DNA fragments.
GeneRuler 100bp Plus DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).
The ladder is supplied with 6X DNA Loading Dye.
The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler 100 bp Plus DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase. The ladder is supplied with 6X DNA Loading Dye.- Ideal for both DNA sizing and approximate quantification
- Sharp bands
- Bright reference bands (given in red)
- Supplied with loading dye for sample DNA
Specifications
| 6X | |
| Xylene Cyanol FF, Bromophenol Blue, Orange G | |
| 100 to 3000 bp | |
| 5 x 50 μg | |
| DNA Ladder |
| Agarose | |
| No | |
| Agarose Gels | |
| GeneRuler |
For Research Use Only. Not for use in diagnostic procedures.