missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Plant RNA Purification Kit

Catalog No. FERK0801
Change view
Click to view available options
Quantity:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0801 50 Preps
FERK0802 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK0801 Supplier Thermo Scientific™ Supplier No. K0801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Requires

Dithiothreitol (DTT, #R0861)

GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.

Thermo Scientific GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.

The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis. Purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis. RNA yields vary between different species, tissues, and age of tissue sample. See Table 1 for typical total RNA yields from various sources.

Highlights

  • Fast—procedure completed in 20 min following cell lysis
  • Convinient—protocol does not include clogging-prone lysate filtration step
  • High yield—up to 65 μg of high purity and high integrity RNA from 50 mg plant sample
  • Compatible with wide variety of samples (leaves, roots, sprouts and other parts of various plants)

Applications

Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:

  • RT-PCR
  • RT-qPCR
  • Northern blotting
  • Nuclease protection assay

Specifications

Content And Storage • 40 mL Plant RNA Lysis Solution; store at room temperature
• 40 mL Wash Buffer WB 1 (concentrated); room temperature
• 30 mL Wash Buffer 2 (concentrated); room temperature
• 30 mL Water, Nuclease-free; room temperature
• 50 GeneJET RNA Spin Columns with Collection Tubes; For better long-term performance, it is recommended to store the purification columns at 2°C-8°C.
• 50 Collection Tubes (2 mL); room temperature
• 50 Collection Tubes (1.5 mL); room temperature

Isolation Technology Silica Spin Column
Purification Time 20 min. (After sample homogenization)
Elution Volume 50 μL
Sample Type Plants
Final Product Type Total RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling
Quantity 50 Preps
Scale Mini
Shipping Condition Room Temperature
Starting Material Amount ≤100 mg
Yield Up to 65 μg
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Is any GeneJET kit suitable for RNA purification from formalin-fixed paraffin-embedded FFPE tissues?

We cannot recommend GeneJET kits for RNA purification from FFPE tissues. In case of genomic DNA purififcation from formalin-fixed paraffin-embedded samples, we recommend GeneJET FFPE DNA Purification Kit.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

Does the GeneJET Stabilized and Fresh Whole Blood RNA Kit isolate miRNA?

This kit is not suitable for purification of small RNAs.

Do the GeneJET RNA purification kits remove genomic DNA?

These kits will remove the majority of genomic DNA, however trace amounts may still remain. Therefore, for applications that are very sensitive to genomic DNA, we recommend you treat the purified RNA sample with DNase I (Cat. No. EN0521), RapidOut DNA Removal Kit (Cat. No. K2981), or dsDNase (Cat. No. EN0771).


For Research Use Only. Not for use in diagnostic procedures.