missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneJET Gel Extraction Kit

Catalog No. FERK0691
Change view
Click to view available options
Quantity:
250 Preps
50 Preps
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERK0691 50 Preps
FERK0692 250 Preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FERK0691 Supplier Thermo Scientific™ Supplier No. K0691
  • ONLINE EXCLUSIVE PRICE
    $169.84
    $107.25 / Each

    Save $62.59

    37.0% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific GeneJET Gel Extraction Kit is designed for rapid and efficient purification of DNA fragments from standard or low-melting point agarose gels run in either TAE or TBE buffer.

Thermo Scientific GeneJET Gel Extraction Kit is designed for rapid and efficient purification of DNA fragments from standard or low-melting point agarose gels run in either TAE or TBE buffer.

The kit utilizes a proprietary silica-based membrane technology in the form of a convenient spin column. The kit can be used to purify DNA fragments from 25 bp to 20 kb in size with recovery rates up to 95%. Each GeneJET purification column has a binding capacity of up to 25 μg of DNA and can process up to 1 g of agarose gel. The entire procedure takes just 15 minutes, and the isolated DNA is ready to use in all common downstream applications, including ligation, restriction digestion, PCR, sequencing, and labeling.

Highlights

  • Highly efficient: up to 95% recoveries in the range of 100 bp to 10 kb
  • Fast: procedure takes only 15 minutes
  • Convenient: spin columns are capped and assembled with collection tubes

Applications

Fast and efficient extraction of high purity DNA fragments ideal for use in all conventional molecular biology procedures including:

  • FastDigest or conventional restriction digestion
  • Automated fluorescent or radioactive sequencing
  • PCR
  • In situ hybridization
  • Labeling
  • Cloning

Specifications

Column Type Spin Column
Content And Storage For better long-term performance, it is recommended to store the purification columns at 2°C to 8°C.
Format Spin Column
Isolation Technology Spin Column
Sample Type Gel
For Use With (Application) Real-Time Quantitative PCR (qPCR), Cloning, Southern Blotting, Nucleic Acid Labeling, Sequencing
High-throughput Compatibility Not High-throughput Compatible (Manual)
Product Line GeneJET
Product Type Gel Extraction Kit
Quantity 50 Preps
Target DNA from Enzymatic Reactions (e.g. PCR)
Test Time 15 min.
Show More Show Less
Can I use water instead of Elution Buffer in the elution step of GeneJET and MagJET nucleic acid purification kits?

We would highly recommend using the provided Elution Buffer in the elution step for best results. You may use nuclease-free water; however, it may affect the yield of the nucleic acids during purification and their stability during storage.

Can I use a vacuum manifold with GeneJET kits?

The GeneJET spin columns are compatible with vacuum manifolds of standard size (e.g. Qiagen, Sigma-Aldrich, Promega). However, not all GeneJET kits are supported with a vacuum protocol. Please refer to the particular kit protocol for the detailed procedure.

Can I use GeneJET Gel Extraction Kit for PCR clean-up?

The GeneJET Gel Extraction Kit is designed for gel extraction but can also be used for PCR clean-up by following the protocol from the GeneJET PCR Purification Kit. The components in the GeneJET Gel Extraction Kit and GeneJET PCR Purification Kit are very similar and can be used interchangeably. The exact composition of solutions in the kits is proprietary.

For a 3-in-1 kit for purification of PCR products from enzymatic reaction mixtures and agarose gels, we recommend using the GeneJET Gel Extraction and DNA Cleanup Micro Kit (Cat. Nos. K0831/2).

At what temperature should I perform centrifugation steps in the GeneJET kits?

We recommend performing centrifugation steps at room temperature when the temperature is not specified.

Are GeneJET spin columns or MagJET magnetic beads available separately?

No, we do not offer GeneJET columns or MagJET magnetic beads separately at this time.

Do you offer the GeneJET Purification Columns included in the GeneJET Gel Extraction Kit as a stand-alone product?

The wash buffers and bindings buffers for the GeneJet kits are in most cases available separately. Unfortunately, the purification columns for all GeneJet kits including the GeneJET Gel Extraction Kit are not available separately. They can only be acquired as part of the whole kit itself.


For Research Use Only. Not for use in diagnostic procedures.