missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FspAI (5 U/μL)

The FspAI restriction enzyme recognizes RTGC^GCAY sites and cuts best at 37°C in O buffer.

Supplier:  Thermo Scientific™ ER1661

 View more versions of this product

Catalog No. FERER1661


Only null left
Explore available promotions

Description

Description

Lambda DNA digested with FspAI, 0.5% agarose, 2 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

More than 80% of the digested DNA fragments can be ligated and more than 90% can be recut at a 5ft.-termini concentration of 0.2μM after a 50-fold overdigestion (3U/μg DNA x 17 hours) with FspAI.

5'...R T G C▵G C A Y...3'

3'...Y A C G▵C G T R...5'

Conditions for 100% Activity:

  • 1X Buffer O: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • FspAI is supplied in: 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Effects:

  • Dam: never overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: completely overlaps — blocked
  • EcoKI: may overlap — effect not determined
  • EcoBI: may overlap — effect not determined

Note:

Low salt, high glycerol (>5%) concentrations, pH >8.0 or a large excess of enzyme may result in star activity.

Specifications

Specifications

5 U/μL
FspAI
Yes
No
Restriction Enzyme
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer O
37°C
100 units
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.