missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eco47III (AfeI) (10 U/μL)

The Eco47III (AfeI) restriction enzyme recognizes AGC^GCT sites and cuts best at 37°C in O buffer (Isoschizomers: AfeI, Aor51HI).

Supplier:  Thermo Scientific™ ER0321

 View more versions of this product

Catalog No. FERER0321


Only null left
Explore available promotions

Description

Description

Lambda DNA digested with Eco47III (AfeI), 0.7% agarose, 2 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...A G C▵G C T...3'

3'...T C G▵C G A...5'

Conditions for 100% Activity:

  • 1X Buffer O:50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • Eco47III is supplied in:10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with Eco47III, more than 80% of pBR322 DNA fragments can be ligated and more than 90% of these can be recut

Methylation Effects:

  • Dam: never overlaps-no effect
  • Dcm: never overlaps-no effect
  • CpG: completely overlaps-blocked
  • EcoKI: never overlaps-no effect
  • EcoBI: may overlap-effect not determined

Digestion of Agarose-embedded DNA:

  • Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours
Specifications

Specifications

10 U/μL
Eco47III (AfeI)
Yes
No
Restriction Enzyme
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer O
37°C
200 units
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.