missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Eco31I (BsaI) (10 U/μL)

The Eco31I (BsaI) restriction enzyme recognizes GGTCTC(1/5)^ sites and cuts best at 37°C in G buffer (Isoschizomers: BsaI, Bso31I, BspTNI).

Supplier:  Thermo Scientific™ ER0291

 View more versions of this product

Catalog No. FERER0291

Only null left

Description

Description

Lambda DNA (dcm-) digested with Eco31I (BsaI), 0.7% agarose, 2 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...G G T C T C (N)1▵...3'

3'...C C A G A G (N)5▵...5'

Conditions for 100% Activity:

  • 1X Buffer G:10mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 50mM NaCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • Eco31I is supplied in:10mM Tris-HCl (pH 7.5 at 25°C), 200mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with Eco31I, more than 95% of DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: never overlaps-no effect
  • Dcm: may overlap-cleavage impaired
  • CpG: may overlap-cleavage impaired
  • EcoKI: never overlaps-no effect
  • EcoBI: may overlap-effect not determined

Digestion of Agarose-embedded DNA:

  • Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Note:

Assayed using lambda DNA (dcm-) (#SD0021), as one of the two Eco31I recognition sites in lambda DNA is difficult to cleave. Eco31I cleavage is impaired by overlappingdcm methylation. To avoiddcm methylation, use adam,dcm- strain such as GM2163 (#M0099). Low salt, high glycerol (>5%) concentrations, pH >8.0 or a large excess of enzyme may result in star activity. Eco31I cleaves downstream of its recognition site and can generate any desired 4 base 5'-overhangs. This feature is useful for direct PCR product cloning.

Specifications

Chemical Identifiers

10 U/μL
Eco31I (BsaI)
Yes
Yes
Scarless Cloning
CpG Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dam Methylation-Sensitive
10X Buffer G
37°C
Restriction Enzyme

Specifications

10 U/μL
Eco31I (BsaI)
Yes
Yes
Restriction Enzyme
CpG Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dam Methylation-Sensitive
10X Buffer G
37°C
1,000 units
Scarless Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.