missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Dilution Buffer for Restriction Enzymes
Optimize applications that require diluted enzymes with this dilution buffer.
Supplier: Thermo Scientific™ B19
Description
Dilution Buffer for Restriction Enzymes is used for applications that require diluted enzymes. Enzymes diluted in this buffer retain 50 to 100% activity after storage for one month at -20°C.
Composition:- 10 mM Tris-HCl (pH 7.4 at 25̡̉C)
- 100 mM KCl
- 1 mM EDTA
- 1 mM DTT
- 50% glycerol
- 0.2 mg/mL BSA
Specifications
| 5 x 1.0 mL | |
| Dilution Buffer | |
| Traditional Cloning |
For Research Use Only. Not for use in diagnostic procedures.