missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ CTP Solution (100 mM)

Catalog No. FERR0451
Change view
Click to view available options
Quantity:
0.25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERR0451 0.25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERR0451 Supplier Thermo Scientific™ Supplier No. R0451

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ CTP (cytidine 5'-triphosphate) is an extremely stable nucleotide with more than 99% purity.

Thermo Scientific™ CTP (cytidine 5'-triphosphate) is an extremely stable nucleotide with more than 99% purity. It is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. This nucleotide is designed for a variety of molecular biology applications.

Features

  • Greater than 99% purity confirmed by HPLC
  • Functionally tested in in vitro transcription
  • Stable for two years at -20°C and after multiple freeze-thaw cycles

Applications

  • aRNA synthesis
  • siRNA synthesis
  • RNA amplification
  • in vitro transcription

Specifications

Concentration 100 mM
For Use With (Application) in vitro Transcription, RNA Amplification
Content And Storage CTP Solution, 100 mM (0.25 mL)

Store at –20°C.
Shipping Condition Dry Ice
Conjugate Unconjugated
Quantity 0.25 mL
Product Type CTP Solution
Form Liquid

For Research Use Only. Not for use in diagnostic procedures.