missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Buffer Set for Restriction Enzymes

Catalog No. FERB30
Change view
Click to view available options
Quantity:
5 x 1.0 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERB30 5 x 1.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERB30 Supplier Thermo Scientific™ Supplier No. B30

Ensure the optimum reaction conditions for each restriction enzyme with this five buffer system.

Our Five Buffer System ensures the optimum reaction conditions for each restriction enzyme. This system consists of 10X B (blue), G (green), O (orange), R (red), and Tango (yellow) buffers. All restriction enzymes are supplied in color-coded tubes to indicate the recommended reaction buffer. The recommended buffer and/or the universal Tango buffer are supplied with each enzyme.

To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation. The buffer set for restriction endonucleases contains 1 mL of each B, G, O, R, and Tango buffers. It is supplied in 10X concentrated stocks.

Thermo Scientific restriction enzymes exhibit 100% of their certified activity in the recommended buffer. However, some enzymes require additives to achieve 100% activity. For example, AjuI, AlfI, BdaI, BplI, BseMII, FaqI, Eco57I, Eco57MI, Hin4I, and TsoI require S-adenosylmethionine, which is supplied with the enzyme, while AarI and BveI require an oligonucleotide (also supplied with the enzyme). Esp3I requires DTT.

The following enzymes require unique buffers for optimal digestion: 

  • AarI
  • AjiI
  • BamHI
  • BfuI
  • Bpu10I
  • BseXI
  • Bsp143I
  • Cfr9I
  • Cfr10I
  • Eam1105I
  • Ecl136II
  • Eco52I
  • EcoRI
  • KpnI
  • Lsp1109I
  • PacI
  • PasI
  • Ppu21I
  • SacI
  • ScaI
  • SdaI
  • SduI
  • SgeI
  • TaqI

Tango buffer has been designed for double digestions of DNA with conventional restriction enzymes.

Specifications

Quantity 5 x 1.0 mL
Product Type Restriction Enzyme
Research Category Traditional Cloning
Can I double digest my DNA using a Thermo Scientific conventional restriction enzyme and a FastDigest restriction enzyme?

For optimal results with fast reaction and 100% buffer compatibility, we highly recommend using FastDigest restriction enzymes in double digestion. In certain cases however, it may be possible to perform double digestion using a mix of Thermo Scientific conventional and Fastdigest restriction enzymes. For specific recommendations, please contact our technical service with detailed information about the enzymes and DNA template you plan to use.

Why do you recommend only 2 µL of 10X Reaction Buffer when digesting unpurified PCR product in a 30 µL reaction?

We recommend only 2 µl 10X Buffer in digestion of unpurified PCR products in 30 ul since salts and ions from the PCR reaction would be carried over to the digestion reaction.

What are key factors promoting star activity?

Star activty may be contributed by:

• Prolonged incubation
• High enzyme concentration
• High glycerol concentration (usually 5% or higher)
• Small reaction volume

Unexpected DNA bands were observed on agarose gel electrophoresis after restriction digestion. What may have caused this?

Unexpected cleavage patterns may be caused by the following reasons:

• Star activity of the restriction enzyme: Make sure to follow the reaction recommendations as specified in the protocol. Star activity may be improved by changing several key factors such as decreasing the reaction time, increasing the reaction volume, and decreasing the enzyme amount.

• Partial or incomplete cleavage (incomplete restriction reaction): Efficiency of the enzyme can be improved by adding more enzyme, prolonging the reaction time, or purifying DNA samples to remove inhibitory contaminants.

• Contamination with non-specific endonucleases: Non-specific endonucleases may be introduced to the DNA sample and/or the enzyme from improper handling, pipetting, etc.

•Improper reaction setup: Mix the digestion reaction thoroughly.

What are possible reasons for incomplete/failed restriction digestion?

The main reason for DNA cleavage reaction failure is the presence of contaminating inhibitors in the template DNA (for example: phenol, chloroform, detergents, ethanol, excess salts, EDTA, etc.). The best way to troubleshoot is to perform control reactions:

1) negative control (experimental DNA in the reaction buffer without the restriction enzyme) to access degradation of DNA by contaminants in the DNA template and/or reaction buffer
2) positive control reaction I (digestion of highly pure control DNA with the restriction enzyme) to access reaction conditions and enzyme activity
3) positive control reaction II (highly pure control DNA + experimental DNA + Restriction Enzyme) to access possible issues with the experimental DNA.

In addition, please check for sensitivity of the restriction enzymes to template DNA methylation.

What is the optimal DNA concentration in restriction enzyme digestion?

The optimal range of DNA concentration is 0.02-0.1 µg/µL in the restriction digestion mixture. We would recommend the volume of the DNA sample not to exceed 30% of the total reaction volume to avoid potential reaction inhibition. To resolve inhibition from impurities of the DNA solution, we recommend increasing the overall volume of the reaction while keeping the volume of the DNA solution the same.

Is there any other way to inactivate Thermo Scientific restriction enzymes (when thermal inactivation is not applicable) without using phenol/chloroform?

To remove the enzymes without using phenol/chloroform, we would recommend silica column based purification such as GeneJET Gel Extraction and DNA Cleanup Micro Kit (Cat. Nos. K0831/2) or GeneJET PCR Purification Kit (Cat. Nos. K0701/2).

What is the stability of Thermo Scientific restriction enzymes if they freeze-thaw during shipment?

Enzymes may freeze during shipment on dry ice. This does not affect their quality as all Thermo Scientific enzymes are tested 100% active after at least three freeze-thaw cycles. For 24-48 hour delivery, enzymes may be shipped on blue ice because their quality is not affected by short exposure to 4 degrees C.

Do Thermo Scientific restriction enzymes come with the buffer(s)?

Thermo Scientific restriction enzymes are shipped with their optimal digestion buffer(s). The buffers are available separately for purchase as well.

What are the best conditions for double digests with conventional Thermo Scientific restriction endonucleases?

First, please note that we changed our restriction enzymes and buffer formulations in 2010. The REact buffers 1-10 were discontinued in favor of a smaller group of universal buffers: B, G, O, R and Tango (Y). The new buffers are not compatible with older restriction enzymes, and it is not recommended to do a double digest with an old enzyme (with REact buffer) and a new enzyme.

When performing any double digest, there may be buffer incompatibilities and enzyme steric hindrance problems. These can be avoided by performing sequential digests, separated by buffer exchange or chloroform extraction and ethanol precipitation. However, if these issues are understood and a double digest will be performed, you can evaluate enzyme combinations using our buffer compatibility chart: https://www.thermofisher.com/us/en/home/brands/thermo-scientific/molecular-biology/thermo-scientific-restriction-modifying-enzymes/restriction-enzymes-thermo-scientific/conventional-restriction-enzymes-thermo-scientific/reaction-conditions-for-restriction-enzymes.html.


For Research Use Only. Not for use in diagnostic procedures.