missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bsp143I (Sau3AI) (10 U/μL)

Cut at ^GATC sites with Bsp143I (Sau3AI), which performs best at 37°C in its own unique buffer (Isoschizomers: BfuCI, BssMI, BstKTI, BstMBI, DpnII, Kzo9I, NdeII, Sau3AIm).

Supplier:  Thermo Scientific™ ER0781

 View more versions of this product

Catalog No. FERER0781

Only null left
Explore available promotions

Description

Description

Conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...▵G A T C ...3'

3'... C T A G▵...5'

Conditions for 100% Activity:

  • 1X Buffer Bsp143I: 33mM Tris-acetate (pH 7.9 at 37°C), 10mM Mg-acetate, 66mM K-acetate, 0.02% Triton X-100 and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • Bsp143I is supplied in: 10mM Tris-HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Ligation and Recleavage:

  • After 50-fold overdigestion with Bsp143I more than 95% of the DNA fragments can be ligated and recut

Methylation Effects:

  • Dam: completely overlaps — no effect
  • Dcm: never overlaps — no effect
  • CpG: may overlap — blocked
  • EcoKI: never overlaps — no effect
  • EcoBI: may overlap — no effect

Note:

DpnI, Bsp143I and MboI all recognize the same sequence but have different methylation sensitivities and cleavage sites. Compatible Ends: BamHI, BclI, BglII, PsuI.

Specifications

Specifications

10 U/μL
Bsp143I (Sau3AI)
Yes
No
Restriction Enzyme
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Unique Buffer (10X Buffer Bsp143I (Sau3AI))
37°C
300 units
Traditional Cloning
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.