missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ATP Solution (100 mM)

Thermo Scientific ATP (adenosine 5'-triphosphate) is an extremely stable nucleotide, and it is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH.

Supplier:  Thermo Scientific™ R0441

Catalog No. FERR0441


Only null left
Explore available promotions

Description

Description

Thermo Scientific ATP (adenosine 5'-triphosphate) is an extremely stable nucleotide, and it is supplied as 100 mM aqueous solution titrated to pH 7.3-7.5 with NaOH. This nucleotide is used in different molecular biology applications, and it has greater than 99% purity.

Features
• Greater than 99% purity confirmed by HPLC
• Functionally tested in in vitro transcription
• Stable for two years at -20°C and after multiple freeze-thaw cycles

Applications
in vitro transcription
• aRNA synthesis
• siRNA synthesis
• RNA amplification
• Ligation
• Phosphorylation

Specifications

Specifications

100 mM
ATP Solution, 100 mM (0.25 mL)

Store at –20°C.
Unconjugated
ATP Solution
in vitro Transcription, RNA Amplification
Dry Ice
0.25 mL
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.