Learn More
Thermo Scientific™ Buffer B (10X)
Ensure the optimum reaction conditions for restriction enzymes with Thermo Scientific™ 10X Buffer B, premixed with BSA for enhanced stability.
Supplier:  Thermo Scientific™ BB5
Catalog No. FERBB5
Only null left
Description
Description
Five Buffer System ensures the optimum reaction conditions for each restriction enzyme. The system consists of 10X B (blue), G (green), O (orange), R (red), and Tango (yellow) buffers. All restriction enzymes are supplied in color-coded tubes to indicate the recommended reaction buffer. The recommended buffer and/or the universal Tango buffer are supplied with each enzyme.
To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.
Restriction enzymes exhibit 100% of their certified activity in the recommended buffer. However, some enzymes require additives to achieve 100% activity. For example, AjuI, AlfI, BdaI, BplI, BseMII, FaqI, Eco57I, Eco57MI, Hin4I, and TsoI require S-adenosylmethionine, which is supplied with the enzyme. AarI and BveI require an oligonucleotide (also supplied with the enzyme), and Esp3I requires DTT.
Specifications
Specifications
10X
5 x 1.0 mL
Buffer B
Traditional Cloning
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.