missing translation for 'onlineSavingsMsg'
Learn More

Fenticonazole nitrate, Thermo Scientific™

Catalog No. p-7238580
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468792500 250 mg
AC468790010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468792500 Supplier Thermo Scientific Chemicals Supplier No. 468792500
Only null left

Chemical Identifiers

CAS 73151-29-8
Molecular Formula C24H21Cl2N3O4S
Molecular Weight (g/mol) 518.41
MDL Number 00941391
InChI Key FJNRUWDGCVDXLU-UHFFFAOYNA-N
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-{[4-(phenylsulfanyl)phenyl]methoxy}ethyl]-1H-imidazole; nitric acid
SMILES O[N+]([O-])=O.ClC1=CC=C(C(CN2C=CN=C2)OCC2=CC=C(SC3=CC=CC=C3)C=C2)C(Cl)=C1

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Crystalline Powder
Color White
Melting Point 133°C to 137°C
Merck Index 13-4033
Quantity 250 mg
Chemical Name or Material Fenticonazole nitrate
Hazard Category Exclamation mark
Hazard Statement Health hazard
EINECSNumber 277-302-6
RTECSNumber NI4780000
Recommended Storage Normal conditions