missing translation for 'onlineSavingsMsg'
Learn More

Fenticonazole Nitrate 97.0+%, TCI America™

Catalog No. F1008200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F1008200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F1008200MG Supplier TCI America Supplier No. F1008200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 73151-29-8
Molecular Formula C24H21Cl2N3O4S
Molecular Weight (g/mol) 518.41
MDL Number MFCD00941391
InChI Key FJNRUWDGCVDXLU-UHFFFAOYNA-N
PubChem CID 51754
ChEBI CHEBI:83606
IUPAC Name 1-[2-(2,4-dichlorophenyl)-2-{[4-(phenylsulfanyl)phenyl]methoxy}ethyl]-1H-imidazole; nitric acid
SMILES O[N+]([O-])=O.ClC1=CC=C(C(CN2C=CN=C2)OCC2=CC=C(SC3=CC=CC=C3)C=C2)C(Cl)=C1

Specifications

Melting Point 135°C
Color White
Quantity 200 mg
Formula Weight 518.41
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Fenticonazole Nitrate
RTECSNumber NI4780000
TSCA No