missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenofibric acid, ≥98.0% (HPLC), MilliporeSigma™ Supelco™
Description
Pharmaceutical (small molecule)
Specifications
Specifications
| CAS | 42017-89-0 |
| Physical Form | Neat |
| Linear Formula | C17H15 ClO4 |
| MDL Number | MFCD00792461 |
| Synonym | 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropionic acid; Fenofibric acid |
| Type | Analytical Standard |
| Quantity | 10 mg |
| Formula Weight | 318.75 |
| Percent Purity | ≥98.0% (HPLC) |
| Grade | Analytical Standard |