missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Fenofibric acid

Catalog No. AC466170010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC466170010 1 g
AC466170050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC466170010 Supplier Thermo Scientific™ Supplier No. 466170010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 42017-89-0
Chemical Name or Material Fenofibric acid

Specifications

Quantity 1 g
Solubility Solubility aq. soln.: .. Other solubilities: practically insoluble in chloroform, soluble in acetone and methanol
Melting Point 187.0°C
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Harmful if swallowed.

Toxic to aquatic life with long lasting effects.

Precautionary Statement GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Avoid release to the environment.

Collect spillage.

Dispose of contents/container to an approved waste disposal plant.

EINECSNumber 255-626-9

RUO – Research Use Only