missing translation for 'onlineSavingsMsg'
Learn More

Fenofibrate 98.0+%, TCI America™

Catalog No. p-7129826
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F06745G 5 g
F067425G 25 g
F0674100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. F06745G Supplier TCI America Supplier No. F06745G

Chemical Identifiers

CAS 49562-28-9
Molecular Formula C20H21ClO4
Molecular Weight (g/mol) 360.834
MDL Number MFCD00133314
InChI Key YMTINGFKWWXKFG-UHFFFAOYSA-N
Synonym fenofibrate, procetofen, antara, lipanthyl, lipantil, tricor, fenobrate, lipidil, secalip, fenoglide
PubChem CID 3339
ChEBI CHEBI:5001
IUPAC Name propan-2-yl 2-[4-(4-chlorobenzoyl)phenoxy]-2-methylpropanoate
SMILES CC(C)OC(=O)C(C)(C)OC1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)Cl

Specifications

Melting Point 81°C
Color White
Quantity 5 g
Formula Weight 360.83
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Fenofibrate
RTECSNumber UA2453400
TSCA No