missing translation for 'onlineSavingsMsg'
Learn More

Fenofibrate Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014319
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014319 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014319 Supplier Merck Emd Millipore Supplier No. PHR1246500MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.Fenofibrate is a third-generation fibric acid derivative which is widely employed for the treatment of atherogenic dyslipidemia, hypertriglyceridemia, mixed dyslipidemia and hypercholesterolemia. Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 49562-28-9
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C20H21 ClO4
MDL Number MFCD00133314
Synonym 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid isopropyl ester
Type Certified Reference Material
Quantity 500 mg
Formula Weight 360.83
Grade Certified Reference Material