missing translation for 'onlineSavingsMsg'
Learn More

Fenofibrate, ≥99% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111014318
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014318 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014318 Supplier Merck Emd Millipore Supplier No. 55443100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 49562-28-9
Physical Form Neat
Linear Formula C20H21 ClO4
MDL Number MFCD00133314
Synonym 2-[4-(4-Chlorobenzoyl)phenoxy]-2-methylpropanoic acid isopropyl ester
Type Analytical Standard
Quantity 100 mg
Formula Weight 360.83
Percent Purity ≥99% (HPLC)
Grade Analytical Standard