missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenhexamid 98.0+%, TCI America™
$268.43 - $797.81
Chemical Identifiers
| CAS | 126833-17-8 |
|---|---|
| Molecular Formula | C14H17Cl2NO2 |
| Molecular Weight (g/mol) | 302.20 |
| MDL Number | MFCD03095700 |
| InChI Key | VDLGAVXLJYLFDH-UHFFFAOYSA-N |
| Synonym | fenhexamid, elevate, teldor, n-2,3-dichloro-4-hydroxyphenyl-1-methylcyclohexanecarboxamide, fenhexamide, decree, fenhexamid iso:bsi, unii-q68c3c9p1u, cyclohexanecarboxamide, n-2,3-dichloro-4-hydroxyphenyl-1-methyl, n-2,3-dichloro-4-hydroxyphenyl-1-methylcyclohexane-1-carboxamide |
| PubChem CID | 213031 |
| ChEBI | CHEBI:81853 |
| IUPAC Name | N-(2,3-dichloro-4-hydroxyphenyl)-1-methylcyclohexane-1-carboxamide |
| SMILES | CC1(CCCCC1)C(=O)NC1=CC=C(O)C(Cl)=C1Cl |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
F0967200MG
|
TCI America
F0967200MG |
200 mg |
Each for $268.43
|
|
|||||
|
F09671G
|
TCI America
F09671G |
1 g |
Each for $797.81
|
|
|||||
Chemical Identifiers
| 126833-17-8 | |
| 302.20 | |
| VDLGAVXLJYLFDH-UHFFFAOYSA-N | |
| 213031 | |
| N-(2,3-dichloro-4-hydroxyphenyl)-1-methylcyclohexane-1-carboxamide |
| C14H17Cl2NO2 | |
| MFCD03095700 | |
| fenhexamid, elevate, teldor, n-2,3-dichloro-4-hydroxyphenyl-1-methylcyclohexanecarboxamide, fenhexamide, decree, fenhexamid iso:bsi, unii-q68c3c9p1u, cyclohexanecarboxamide, n-2,3-dichloro-4-hydroxyphenyl-1-methyl, n-2,3-dichloro-4-hydroxyphenyl-1-methylcyclohexane-1-carboxamide | |
| CHEBI:81853 | |
| CC1(CCCCC1)C(=O)NC1=CC=C(O)C(Cl)=C1Cl |
Specifications
| 126833-17-8 | |
| 320°C | |
| MFCD03095700 | |
| 3077 | |
| VDLGAVXLJYLFDH-UHFFFAOYSA-N | |
| N-(2,3-dichloro-4-hydroxyphenyl)-1-methylcyclohexane-1-carboxamide | |
| 213031 | |
| 302.20 | |
| Crystalline Powder |
| 153°C | |
| C14H17Cl2NO2 | |
| 200 mg | |
| fenhexamid, elevate, teldor, n-2,3-dichloro-4-hydroxyphenyl-1-methylcyclohexanecarboxamide, fenhexamide, decree, fenhexamid iso:bsi, unii-q68c3c9p1u, cyclohexanecarboxamide, n-2,3-dichloro-4-hydroxyphenyl-1-methyl, n-2,3-dichloro-4-hydroxyphenyl-1-methylcyclohexane-1-carboxamide | |
| CC1(CCCCC1)C(=O)NC1=CC=C(O)C(Cl)=C1Cl | |
| 302.20 | |
| CHEBI:81853 | |
| ≥98.0% (HPLC) | |
| Fenhexamid |
Safety and Handling
RTECSNumber : GU7879550
TSCA : No