missing translation for 'onlineSavingsMsg'
Learn More

(-)-Fenchone, ≥97% (GC), MilliporeSigma™ Supelco™

Catalog No. 111019761
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019761 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019761 Supplier Merck Emd Millipore Supplier No. 9210150MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Food and Beverages

Specifications

CAS 7787-20-4
Density 0.948 g/mL (at 25°C)
Linear Formula C10H16 O
MDL Number MFCD00151104
Melting Point 5°C to 6°C
Boiling Point 192°C to 194°C
Synonym (1 R)-(-)-Fenchone; (-)-1,3,3-Trimethyl-2-norbornanone; (-)-Fenchone; (1 R)-1,3,3-Trimethylbicyclo[2.2.1 ]heptan-2-one
Type Analytical Standard
Quantity 50 mg
Formula Weight 152.23
Percent Purity ≥97% (GC)
Grade Analytical Standard
Show More Show Less