missing translation for 'onlineSavingsMsg'
Learn More

BTX™ Female/Female Adapter Set

Catalog No. BTX450088
Change view
Click to view available options
Connection Type:
Banana Cables
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Connection Type
BTX450088 Banana Cables
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BTX450088 Supplier BTX™ Supplier No. 450088

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Female/Female Adapter Set for 4 mm diameter Banana Plug Cables.

  • For connection to electroporation generators
  • 4 mm Female/Female Adapter Set

Specifications

For Use With (Application) To connect an electrofusion/electroporation system to the electrodes and chambers to be used for a particular application
For Use With (Equipment) 4 mm dia. Banana Plug Cables 45-0216
Product Line BTX Electroporation
Product Type Electroporation accessory
Description Female/Female Adapter Set for Banana Plug Cables 45-0216
Display LCD
Length (English) Cable 10 ft.
Length (Metric) Cable 3.1 m
Sterility Non-sterile
Connection Type Banana Cables
Pulse Form Exponential Decay Wave, Square Wave
Show More Show Less