missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Felodipine | |
| 144°C | |
| C18H19Cl2NO4 | |
| 5 g | |
| RZTAMFZIAATZDJ-UHFFFAOYSA-N | |
| 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate | |
| 3333 | |
| 384.25 | |
| Crystalline Powder |
| 72509-76-3 | |
| Yellow | |
| MFCD00868316 | |
| felodipine, plendil, flodil, munobal, renedil, dl-felodipine, perfudal, splendil, feloday, prevex | |
| CCOC(=O)C1=C(NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C)C | |
| 384.253 | |
| CHEBI:585948 | |
| ≥98.0% (HPLC,N) |
Chemical Identifiers
| 72509-76-3 | |
| 384.253 | |
| RZTAMFZIAATZDJ-UHFFFAOYSA-N | |
| 3333 | |
| 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate |
| C18H19Cl2NO4 | |
| MFCD00868316 | |
| felodipine, plendil, flodil, munobal, renedil, dl-felodipine, perfudal, splendil, feloday, prevex | |
| CHEBI:585948 | |
| CCOC(=O)C1=C(NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C)C |
Safety and Handling
RTECSNumber : US7968700
TSCA : No