missing translation for 'onlineSavingsMsg'
Learn More

Felodipine 98.0+%, TCI America™

Catalog No. F08141G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08141G 1 g
F08145G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08141G Supplier TCI America Supplier No. F08141G

Chemical Identifiers

CAS 72509-76-3
Molecular Formula C18H19Cl2NO4
Molecular Weight (g/mol) 384.253
MDL Number MFCD00868316
InChI Key RZTAMFZIAATZDJ-UHFFFAOYSA-N
Synonym felodipine, plendil, flodil, munobal, renedil, dl-felodipine, perfudal, splendil, feloday, prevex
PubChem CID 3333
ChEBI CHEBI:585948
IUPAC Name 5-O-ethyl 3-O-methyl 4-(2,3-dichlorophenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CCOC(=O)C1=C(NC(=C(C1C2=C(C(=CC=C2)Cl)Cl)C(=O)OC)C)C

Specifications

Melting Point 144°C
Color Yellow
Quantity 1 g
Formula Weight 384.25
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Felodipine
RTECSNumber US7968700
TSCA No