missing translation for 'onlineSavingsMsg'
Learn More

Fehlings Solution 'A', Copper No. 1, Certified, 6.93% (w/v) ±0.30%, LabChem™

Supplier:  LabChem LC142004

 View more versions of this product

Catalog No. LC142004


Only null left

Description

Description

Specifications

Specifications

Fehlings Solution 'A', Copper No. 1
7758-99-8 , 7732-18-5
CuSO4·5H2O
Passes Test
6.93% ±0.3% (w/v)
Poly Bottle
Blue
Liquid
Copper No. 1
93.07,6.93
Soluble in water
249.68
Certified
Sulfur compounds; Fume
4 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions