Learn More
Fehlings Solution 'A', Copper No. 1, Certified, 6.93% (w/v) ±0.30%, LabChem™
Supplier:  LabChem LC142002
 View more versions of this product
Catalog No. LC142002
Only null left
Description
Description
Specifications
Specifications
Fehling's Solution A
7758-99-8 , 7732-18-5
CuSO4·5H2O
6.93% ±0.3% (w/v)
Poly Bottle
Liquid
Copper No. 1
93.07,6.93
249.68
Certified
1 L
Product Suggestions
Product Suggestions