missing translation for 'onlineSavingsMsg'
Learn More

Fehling's Copper Solution (Fehling Solution A), Ricca Chemical

For Determination of Reducing Sugars

$73.47 - $256.90

Specifications

Chemical Name or Material Fehling's Copper Solution
Color Blue
Name Note Fehling Solution A
CAS 7758-99-8,7732-18-5
Grade Laboratory
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
3010-16
View Documents
Ricca Chemical Company
RI301016
500 mL Natural Poly Bottle
Each for $73.47
Only null left
 
3010-32
View Documents
Ricca Chemical Company
RI301032
1 L Natural Poly Bottle
Each for $108.47
Only null left
 
3010-1
View Documents
Ricca Chemical Company
RI30101
4 L Natural Poly Bottle
Each for $256.90
Only null left
 
Description

Description

Specifications

Specifications

Fehling's Copper Solution
Fehling Solution A
Laboratory
Liquid
Blue
7758-99-8,7732-18-5
∼4
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'