missing translation for 'onlineSavingsMsg'
Learn More

Feed Kit A+, B+, C+

Catalog No. A3315801
Change view
Click to view available options
Quantity:
3 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3315801 3 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A3315801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

Content And Storage • 1 x 1 L EfficientFeed A+ Supplement
• 1 x 1 L EfficientFeed B+ Supplement
• 1 x 1 L EfficientFeed C+ Supplement
Store above in refrigerator (2–8°C). Protect from light and moisture.

• Flash drive with manual

Cell Line CHO
Classification Chemically-defined
Form AGT
Product Type Media Supplement
Quantity 3 L