missing translation for 'onlineSavingsMsg'
Learn More

Febuxostat 97.0+%, TCI America™

Catalog No. p-7129937
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F08471G 1 g
F08475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. F08471G Supplier TCI America Supplier No. F08471G
Only null left

Chemical Identifiers

CAS 144060-53-7
Molecular Formula C16H16N2O3S
Molecular Weight (g/mol) 316.38
MDL Number MFCD00871598
InChI Key BQSJTQLCZDPROO-UHFFFAOYSA-N
Synonym febuxostat, adenuric, uloric, feburic, tmx 67, 2-3-cyano-4-isobutoxyphenyl-4-methylthiazole-5-carboxylic acid, tmx-67, febuxostat usan, unii-101v0r1n2e, uloric tn
PubChem CID 134018
ChEBI CHEBI:45943
IUPAC Name 2-[3-cyano-4-(2-methylpropoxy)phenyl]-4-methyl-1,3-thiazole-5-carboxylic acid
SMILES CC(C)COC1=CC=C(C=C1C#N)C1=NC(C)=C(S1)C(O)=O

Specifications

Melting Point 209°C
Color White-Yellow
Quantity 1 g
Formula Weight 316.38
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Febuxostat
RTECSNumber XJ3675310
TSCA No