missing translation for 'onlineSavingsMsg'
Learn More

Febantel, Thermo Scientific™

Catalog No. p-7232410
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468780010 1 g
AC468780050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468780010 Supplier Thermo Scientific Chemicals Supplier No. 468780010
Only null left

Chemical Identifiers

CAS 58306-30-2
Molecular Formula C20H22N4O6S
Molecular Weight (g/mol) 446.48
MDL Number 01738527
InChI Key HMCCXLBXIJMERM-UHFFFAOYSA-N
IUPAC Name methyl N-{[(methoxycarbonyl)imino]({[2-(2-methoxyacetamido)-4-(phenylsulfanyl)phenyl]amino})methyl}carbamate
SMILES COCC(=O)NC1=CC(SC2=CC=CC=C2)=CC=C1NC(NC(=O)OC)=NC(=O)OC

Specifications

Packaging Glass Bottle
Certificate of Analysis Available
Physical Form Powder/Crystals
Color White to Light Yellow
Melting Point 129°C to 130°C
Merck Index 3973
Solubility Insoluble in water
Quantity 1 g
Formula Weight 446.48
Chemical Name or Material Febantel
Hazard Category GHS07: Exclamation mark
ShelfLife 5 years
EINECSNumber 261-205-0
RTECSNumber FB3955000
Recommended Storage Freezer -20°C