missing translation for 'onlineSavingsMsg'
Learn More

Febantel, TCI America™

Catalog No. F09221G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F09221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F09221G Supplier TCI America Supplier No. F09221G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 58306-30-2
Molecular Formula C20H22N4O6S
Molecular Weight (g/mol) 446.48
MDL Number MFCD01738527
InChI Key HMCCXLBXIJMERM-UHFFFAOYSA-N
PubChem CID 9570638
IUPAC Name methyl N-{[(methoxycarbonyl)imino]({[2-(2-methoxyacetamido)-4-(phenylsulfanyl)phenyl]amino})methyl}carbamate
SMILES COCC(=O)NC1=CC(SC2=CC=CC=C2)=CC=C1NC(NC(=O)OC)=NC(=O)OC

Specifications

Melting Point 130°C
Quantity 1 g
Formula Weight 446.48
Chemical Name or Material Febantel
RTECSNumber FB3955000
TSCA No