missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fe65 Recombinant Rabbit Monoclonal Antibody (23GB3665)

Catalog No. MA553449
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553449 20 μL
MA553448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553449 Supplier Invitrogen™ Supplier No. MA553449
Only null left

Rabbit Recombinant Monoclonal Antibody

Fe65 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The protein encoded by this gene is a member of the Fe65 protein family. It is an adaptor protein localized in the nucleus. It interacts with the Alzheimer's disease amyloid precursor protein, transcription factor CP2/LSF/LBP1 and the low-density lipoprotein receptor-related protein. APP functions as a cytosolic anchoring site that can prevent the gene product's nuclear translocation. This encoded protein could play an important role in the pathogenesis of Alzheimer's disease. It is thought to regulate transcription. Also it is observed to block cell cycle progression by downregulating thymidylate synthase expression. Multiple alternatively spliced transcript variants have been described for this gene but some of their full length sequence is not known.

Specifications

Antigen Fe65
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB3665
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene APBB1
Gene Accession No. O00213
Gene Alias adaptor protein FE65a2; amyloid beta (A4) precursor protein-binding, family B, member 1; amyloid beta (A4) precursor protein-binding, family B, member 1 (Fe65); amyloid beta A4 precursor protein-binding family B member 1; amyloid beta precursor protein binding family B member 1; Amyloid-beta A4 precursor protein-binding family B member 1; APBB1; FE65; MGC:9072; Protein Fe65; RIR; stat-like protein
Gene Symbols APBB1
Host Species Rabbit
Immunogen A synthetic peptide of human FE65 (650-710AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 322
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less