Learn More
METTLER TOLEDO FE Standard Precision Portable Balance
Standard FE Precision Portable Balances are highly accurate, efficient, simple to operate, and designed with robust construction for proven long-lasting performance. Intuitive onscreen guidance within the LCD hybrid touchscreen allows you to easily navigate through 9 built-in applications.
Supplier: METTLER TOLEDO 30938248
Description
- External adjustment
- Portable balance
- LCD hybrid touchscreen
- Intuitive onscreen guidance
- 10 built-in applications
- Statistical data analysis
- USB connection
- Metal base with PBT top housing
- Overload protection
- Have a legacy METTLER TOLEDO ME balance? The FE balance is your new solution. See how it ranks among our balance lines:
- XPR Excellence Precision Balances
- MX Advanced Precision Balances
- MR Advanced Precision Balances
- MA Standard Precision Balances
- FE Standard Precision Balances
- LA Standard Precision Balances
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
2200 g | |
0.01 g | |
FE2002PE | |
1 s |
LCD Touchscreen | |
14 g | |
0.01 g | |
Precision Balance |