missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FD and C Blue No. 1, Spectrum™ Chemical
FD110, 3844-45-9, C37H34N2O9S3Na2
$322.71 - $34,850.07
Chemical Identifiers
| CAS | 3844-45-9 |
|---|---|
| Molecular Formula | C37H34N2Na2O9S3 |
| Molecular Weight (g/mol) | 792.84 |
| InChI Key | SGHZXLIDFTYFHQ-UHFFFAOYSA-L |
| IUPAC Name | disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(2-sulfonatophenyl)methylium |
| SMILES | [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=CC=C1S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18602610
|
N/A | 25 g | Fiber Drum |
Each for $322.71
|
|
||||
|
18602606
|
N/A | 100 g | Fiber Drum |
Each for $663.25
|
|
||||
|
18602607
|
N/A | 500 g | Fiber Drum |
Each for $1,614.29
|
|
||||
|
18602609
|
N/A | 10 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18602608
|
N/A | 45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical FD and C Blue No. 1 is a synthetic dye produced from petroleum. It is used as a coloring agent, biological stain, textile dye, wood stain, and indicator. In cosmetics and personal care products, it is used in the formulation of a wide variety of product types, eye makeup, makeup, lipsticks, nail polishes, personal cleansers and skin care products. It appears as a dark violet to reddish violet, crystalline powder that is water soluble. In the United States, the Food and Drug Administration (FDA) controls the specifications for food, drug and cosmetic colors. Spectrum certifies all batches of artificial food colors meet FDA FD&C quality.Chemical Identifiers
| 3844-45-9 | |
| 792.84 | |
| disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(2-sulfonatophenyl)methylium |
| C37H34N2Na2O9S3 | |
| SGHZXLIDFTYFHQ-UHFFFAOYSA-L | |
| [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=CC=C1S([O-])(=O)=O |
Specifications
| 3844-45-9 | |
| C37H34N2Na2O9S3 | |
| [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=CC=C1S([O-])(=O)=O | |
| 792.84 | |
| Reagent | |
| FD and C Blue No. 1 |
| 100% | |
| SGHZXLIDFTYFHQ-UHFFFAOYSA-L | |
| disodium bis(4-{ethyl[(3-sulfonatophenyl)methyl]amino}phenyl)(2-sulfonatophenyl)methylium | |
| 85% | |
| Purple/Violet |