missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ FCGRT Polyclonal Antibody

Catalog No. PIPA597738
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA597738 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA597738 Supplier Invitrogen™ Supplier No. PA597738
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FCGRT Polyclonal Antibody

FCGRT Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

Specifications

Antigen FCGRT
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Gene FCGRT
Gene Accession No. P55899, Q61559
Gene Alias alpha-chain; Fc fragment of IgG receptor and transporter; Fc fragment of IgG, receptor, transporter, alpha; Fc receptor, IgG, alpha chain transporter; FCGRT; FCRN; FcRn alpha chain; heavy chain of the major histocompatibility complex class I-like Fc receptor; IgG Fc fragment receptor transporter alpha chain; IgG receptor FcRn large subunit p51; immunoglobulin receptor, intestinal, heavy chain; major histocompatibility complex class I-like Fc receptor; neonatal Fc receptor; neonatal Fc-receptor for Ig; transmembrane alpha chain of the neonatal receptor
Gene Symbols FCGRT
Host Species Rabbit
Immunogen Recombinant Human IgG receptor FcRn large subunit p51 protein (24-297AA).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14132, 2217
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less