missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human FcGR2BC (CD32 B/C) ELISA Kit

ELISA

Supplier:  Invitrogen™ EHFCGR2B

Encompass

 View more versions of this product

Catalog No. EHFCGR2B

Only null left

Description

Description

The Human FcGR2BC (CD32 B/C) ELISA quantitates Hu FcGR2BC in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu FcGR2BC. Principle of the method The Human FcGR2BC solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

This gene encodes one member of a family of immunoglobulin Fc receptor genes found on the surface of many immune response cells. The protein encoded by this gene is a cell surface receptor found on phagocytic cells such as macrophages and neutrophils, and is involved in the process of phagocytosis and clearing of immune complexes. Alternative splicing results in multiple transcript variants.
Specifications

Chemical Identifiers

P31994,P31995
85 pg/mL
HRP
Other Proteins
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA- 78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL- 11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF- 7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
2212, 2213, 9103
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 2.5 μL, Serum 2.5 μL, Supernatant 100 μL
RP11-474I16.2, CD32, CD32B, FCG2, FCGR2, IGFR2
1 hrs 20 mins
Solid-phase sandwich ELISA
78.13 to 5000 pg/mL
85 pg/mL to 5000 pg/mL
ELISA
Plasma, Serum, Supernatant
Human
Colorimetric Microplate Reader
CD32,CD32B,FCG2,FCGR2,IGFR2,CD32,FCG2,CD32C,CDW32,IGFR2,FCRIIC
<10
HRP
Immunology
2°C to 8°C
CD32
4 hrs 45 mins

Specifications

P31994,P31995
85 pg/mL
Solid-phase sandwich ELISA
ELISA
Plasma, Serum, Supernatant
96 Assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
2212, 2213, 9103
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
96 Tests
Immunology
2°C to 8°C
CD32
4 hrs 45 mins
78.13 to 5000 pg/mL
85 pg/mL to 5000 pg/mL
HRP
Other Proteins
This ELISA kit shows no cross-reactivity with the following cytokines tested: human Angiogenin, BDNF, BLC, CNTF, ENA- 78, FGF-4, IL-1 alpha, IL-1 beta, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL- 11, IL-12 p70, IL-12 p40, IL-13, IL-15, IL-309, IP-10, FGF-4, FGF-6, FGF- 7, G-CSF, GDNF, GM-CSF, IFN-gamma, IGFBP-2, IGFBP-3, IGFBP-4, Leptin (OB), MCP-1, MCP-2, MCP-3, MDC, MIF, MIG, MIP-1 alpha, MIP-1 beta, MIP-1 delta, PARC, PDGF, RANTES, SCF, SDF-1 alpha, TARC, TGF-beta, TIMP-1, TIMP-2, TNF-alpha, TNF-beta, TPO, VEGF.
Human
Colorimetric Microplate Reader
CD32,CD32B,FCG2,FCGR2,IGFR2,CD32,FCG2,CD32C,CDW32,IGFR2,FCRIIC
<10
HRP
RUO
Plasma 2.5 μL, Serum 2.5 μL, Supernatant 100 μL
RP11-474I16.2, CD32, CD32B, FCG2, FCGR2, IGFR2
1 hrs 20 mins
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions