missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FCGBP Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5149779
Description
FCGBP Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| FCGBP | |
| Polyclonal | |
| Unconjugated | |
| FCGBP | |
| Fc fragment of IgG binding protein; FC(GAMMA)BP; Fcgamma-binding protein antigen; fcgammaBP; FCGBP; Human Fc gamma BP; IgG Fc binding protein; IgGFc-binding protein | |
| Rabbit | |
| Protein A | |
| RUO | |
| 8857 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q9Y6R7 | |
| FCGBP | |
| Recombinant Fc Fragment Of IgG Binding Protein (FcgBP) (Cys5176-Ala5344). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |