missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Fc OxyBURST™ Green Assay Reagent

Catalog No. 501137418
Encompass
Change view
Click to view available options
Quantity:
500 μL, 25 reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501137418 500 μL, 25 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501137418 Supplier Invitrogen™ Supplier No. F2902

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Fc OxyBURST™ Green Assay Reagent

The Fc OxyBURST™ Green assay reagent comprises bovine serum albumin (BSA) that has been covalently linked to dichlorodihydrofluorescein (H2DCF) and then complexed with a purified rabbit polyclonal anti-BSA antibody. When this immune complex binds to Fc receptors, the nonfluorescent H2DCF molecules are internalized within the phagovacuoloe and subsequently oxidized to green-fluorescent 2''7'-dichlorofluorescein.

Specifications

Concentration 3 mg⁄ml
Content And Storage Contains 1 vial of Fc OxyBURST™ assay green reagent (suspension in PBS containing sodium azide). Store in refrigerator (2–8°C) and protect from light.
Detection Method Fluorescence
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Assay Reagent
Dye Type Free Radical Sensor
Form Solution
Format Tube(s), Slide(s)
Product Line OXYBURST
Quantity 500 μL, 25 reactions
Shipping Condition Wet Ice
Solubility DMSO (Dimethylsulfoxide)
Emission Wavelength 488/523
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.