missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FBXW11 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576489
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbws class and, in addition to an F-box, contains multiple WD40 repeats. This gene contains at least 14 exons, and its alternative splicing generates 3 transcript variants diverging at the presence/absence of two alternate exons.Specifications
| FBXW11 | |
| Polyclonal | |
| Unconjugated | |
| Fbxw11 | |
| 2310065A07Rik; AA536858; beta-transducin repeat-containing protein 2; BTRC2; BTRCP2; F-box and WD repeat domain containing 11; F-box and WD repeats protein beta-TrCP2; F-box and WD-40 domain protein 11; F-box and WD-40 domain protein 1B; F-box protein Fbw1b; F-box/WD repeat-containing protein 11; F-box/WD repeat-containing protein 1B; Fbw11; FBW1B; FBXW11; Fbxw1b; homologous to Slimb protein; HOS; hypothetical protein; KIAA0696 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23291 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9UKB1 | |
| Fbxw11 | |
| Recombinant fusion protein of human FBXW11. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |