missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ FAXC Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558657
Description
Immunogen sequence: HFYWTLAYCQ WVDNLNETRK MLSLSGGGPF SNLLRWVVCH ITKGIVKREM HGHGIGRFSE EEIYMLMEKD MRSLAGLLG Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Chemical Identifiers
| FAXC | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| FAXC | |
| 6230409E13Rik; AW742931; C6orf168; dJ273F20; Failed axon connections homolog; failed axon connections homolog (Drosophila); FAXC; RGD1562449 | |
| FAXC | |
| Primary | |
| Antigen affinity chromatography |
| 0.1 mg/mL | |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| D3ZAT9, Q3UMF9, Q5TGI0 | |
| 366333, 76132, 84553 | |
| Recombinant protein corresponding to Human FAXC. Recombinant protein control fragment (Product #RP-94887). | |
| Antibody |
Specifications
| FAXC | |
| Polyclonal | |
| Unconjugated | |
| FAXC | |
| 6230409E13Rik; AW742931; C6orf168; dJ273F20; Failed axon connections homolog; failed axon connections homolog (Drosophila); FAXC; RGD1562449 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 366333, 76132, 84553 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| D3ZAT9, Q3UMF9, Q5TGI0 | |
| FAXC | |
| Recombinant protein corresponding to Human FAXC. Recombinant protein control fragment (Product #RP-94887). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |