Learn More
Faucet-Mount Eyewashes
Eyewashes attach easily to any standard laboratory faucet; no special plumbing needed.
Supplier: Customer Channels Group (Ccg) EPDEW100
Alert:
To avoid scalding, cold water or tempered water connection is recommended
Includes:
Common adapters that fit most standard and gooseneck faucets
Description
- Made of chrome-plated brass with float-off dust covers
- Back-mount unit provides 1.5 gpm
- Angled unit has flow control
Compliance
Meet ANSI Z358.1-1998 standards
Specifications
| ANSI Z358.1-1998 | |
| Eyewash | |
| Back |
| 1.5 gpm | |
| Float-off dust covers, diverter, Chrome Plated Brass valve, adapters | |
| Opti-Klensie Eyewash |