missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FastVac™ Vacuum Manifold

Catalog No. A35899
Encompass
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A35899 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A35899 Supplier Thermo Scientific™ Supplier No. A35899

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The one-way Luer-Lock stopcocks are compatible with most nucleic acid purification columns and individually controlled, allowing the processing of as few as one sample at a time.

The one-way Luer-Lock stopcocks are compatible with most nucleic acid purification columns and individually controlled, allowing the processing of as few as one sample at a time. Furthermore, the extra-large waste reservoir makes the FastVac Vacuum Manifold convenient for protocols that require the processing of large volumes, such as during plasmid DNA isolation using a PureLink Fast Low-Endotoxin plasmid purification kit.

Product features:
• Capacity—capable of processing 1 to 20 samples simultaneously using columns and a vacuum source
• Composition—consists of a linear, low-density polyethylene manifold body, one-way Luer-Lock stopcocks made of a polycarbonate body and polypropylene handle, a polypropylene vacuum hose connector, and a neoprene stopper
• Compatibility—compatible with most nucleic acid purification kits (refer to the kit instruction manual for information regarding compatibility and processing with a vacuum manifold)

Specifications

Product Type Vacuum Manifold
Content And Storage • 1 vacuum manifold
• 20 Luer-Lock connectors
• 1 Neoprene stopper

Product Line FastVac
Quantity 1 Ea.
Shipping Condition Room Temperature

For Research Use Only. Not for use in diagnostic procedures.