Learn More
Thermo Scientific™ FastRuler DNA Ladders, ready-to-use
Thermo Scientific FastRuler DNA Ladders are engineered for fast and convenient sizing and approximate quantification of double stranded DNA fragments.
Supplier:  Thermo Scientific™ SM1113
 View more versions of this product
Catalog No. FERSM1113
Only null left
Explore available promotions
Description
Description
Thermo Scientific FastRuler DNA Ladders is a fast and convenient tool for double-stranded DNA sizing during electrophoresis. It's specifically engineered for rapid sizing and approximate quantification of DNA on both conventional agarose gels and high-throughput E-Gels (in 48- and 96-well formats).
The ladder is a mixture of five blunt-ended chromatography-purified individual DNA fragments. This allows you to achieve precise results faster, saving valuable lab time.
Highlights
  • Variety of available ranges (from ultra-low to high- range)
  • Fast separation (8 to 14 min)
  • Short separation distance (10 to 20 mm)
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready-to-use: premixed with 6X MassRuler DNA Loading Dye for direct loading
  • Stable at room temperature for up to 24 months
Sustainability features
Ready-to-use FastRuler DNA ladders can be stored at room temperature for up to 24 months. The ladder is shipped at ambient temperature, using optimized right-sized packaging and 100% recyclable packaging reducing waste.
Specifications
Specifications
Agarose
Middle Range DNA Ladder
100 to 5000 bp
Sustainable packaging
1000 μL
Bromophenol Blue
Yes
Agarose Gels
FastRuler
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.