missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ FastCellular™ Senescence Detection Kit, SPiDER-ΒGal

Detect senescent cells with much higher sensitivity than X-gal.

Supplier:  MP Biomedicals™ 092690301

Catalog No. MP092690301


Only null left
Explore available promotions

Description

Description

Cellular senescence is a phenomenon by which normal cells cease to divide and enter a state of irreversible growth arrest. It is one of the most fundamental aspects of cellular behavior and generally believed to be associated with tumor suppressive mechanisms and an underlying cause of aging. Senescence-associated Β-galactosidase (SA-Β-gal), which is overexpressed in senescent cells, has been widely used as a marker of cellular senescence. Although X-gal is a well-known reagent used to detect SA-Β-gal, X-gal has the following disadvantages: 1) requirement of fixed cells due to poor cell-permeability, 2) low quantitative capability because of the difficulty in visually differentiating stained vs. non-stained cells 3) requirement for long stain times.

To conquer the disadvantages inherent in using X-gal, we utilize a new reagent and method for the detection of cellular senescence. SPiDER-ΒGal offers increased sensitivity in detecting SA-Β-gal due to increased cell permeability and retention inside cells. Compared to the X-gal method, the FastCellular Senescence Detection Kit can be used for both living and fixed cells, because we provide Bafilomycin A1 to specifically inhibit endogenous Β-galactosidase activity in living cells and thus increase the detection specificity for living cells. For fixed cells, SA-Β-gal is detectable by using SPiDER-ΒGal and Mcllvaine buffer (pH 6.0, please refer to the protocol for fixed cell assays). Since SPiDER-ΒGal emits strong and stable fluorescence after the reaction with SA-Β-gal, it can be applied to quantitative analysis by flow cytometry. In addition, the staining time is only 30 mins. Kit includes a 6-well plate with 1 unit of SPiDER-ΒGal and 1 unit of Bafilomycin A1

Benefits of using FastCellular Senescence Detection Kit:

  • Higher sensitivity due to a new fluorogenic detection probe
  • Applicable for living cells and fixed tissues
  • Ability to quantify SA-Β-gal in cellular senescence assay
  • Short staining time (30 min)
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
Specifications

Specifications

Store at 0 to 5°C with protection from light
Cellular Senescence Detection
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions