Learn More
GVS FAST™ Slide Incubation Chambers
Desgned with gasket to form tight and leak-proof seal
$984.97 - $1048.97
Specifications
| Type | Incubation Chamber |
|---|---|
| For Use With (Equipment) | FAST™ Slides |
| Quantity | 10/Pk. |
| Catalog Number | Mfr. No. | Format | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Format | Price | Quantity | |||||
|
10-486-087
|
GVS
10486087 |
2-well |
Pack of 10 Each for $1,048.97
|
|
|||||
|
10-486-137
|
GVS
10486137 |
16-array Plate |
Pack of 10 Each for $984.97
|
|
|||||
Description
- Used in conjunction with FAST Frame or Chip Clip™ Slide Holder
- Conducts binding reactions on protein microarrays
- Designed to fit all FAST Slide formats (1-, 2-, 8- and 16-pad)
- With sensitivity and dynamic range
- Provides long-term stability of printed proteins
- Compatible with all detection methodologies
- Compatible with commercially available arraying robots
Specifications
| Incubation Chamber | |
| FAST™ Slides | |
| 10/Pk. |