Learn More
GVS FAST™ Slide Incubation Chambers
Desgned with gasket to form tight and leak-proof seal
Supplier:  GVS 10486137
 View more versions of this product
Catalog No. 10-486-137
Only null left
Explore available promotions
Description
Description
  • Used in conjunction with FAST Frame or Chip Clip™ Slide Holder
  • Conducts binding reactions on protein microarrays
  • Designed to fit all FAST Slide formats (1-, 2-, 8- and 16-pad)
  • With sensitivity and dynamic range
  • Provides long-term stability of printed proteins
  • Compatible with all detection methodologies
  • Compatible with commercially available arraying robots
Specifications
Specifications
Incubation Chamber
16-array Plate
FAST™ Slides
10/Pk.
Product Suggestions
Product Suggestions