missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Green FCF, pure, certified

CAS: 2353-45-9 | C37H34N2Na2O10S3 | 808.84 g/mol

Supplier:  Thermo Scientific Chemicals 410530250

 View more versions of this product

Catalog No. AC410530250

Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Fast Green FCF
2353-45-9
85% min.
MFCD00013053
Solubility in water: 150g/L (25°C).
Authentic
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
16887
808.84
Glass bottle
25 g
290.0°C
0.98 to 1.2
C37H34N2Na2O10S3
C.I. 42053
XJBPDZVCYTYRIN-UHFFFAOYSA-L
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
808.84
622 to 626nm
Pure
Bordeaux to Maroon
Powder

Chemical Identifiers

2353-45-9
808.84
XJBPDZVCYTYRIN-UHFFFAOYSA-L
16887
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
C37H34N2Na2O10S3
MFCD00013053
C.I. 42053
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only