missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Fast Green FCF, pure, certified

CAS: 2353-45-9 | C37H34N2Na2O10S3 | 808.84 g/mol

$303.77 - $866.42

Chemical Identifiers

CAS 2353-45-9
Molecular Formula C37H34N2Na2O10S3
Molecular Weight (g/mol) 808.84
MDL Number MFCD00013053
InChI Key XJBPDZVCYTYRIN-UHFFFAOYSA-L
Synonym C.I. 42053
PubChem CID 16887
IUPAC Name disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
SMILES [Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC410530250
View Documents Promotion Details
Thermo Scientific Chemicals
410530250
25 g Glass bottle
Each for $303.77
Only null left
 
AC410531000
View Documents Promotion Details
Thermo Scientific Chemicals
410531000
100 g Glass bottle
Each for $866.42
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2353-45-9
808.84
XJBPDZVCYTYRIN-UHFFFAOYSA-L
16887
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
C37H34N2Na2O10S3
MFCD00013053
C.I. 42053
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate

Specifications

290.0°C
0.98 to 1.2
C37H34N2Na2O10S3
C.I. 42053
XJBPDZVCYTYRIN-UHFFFAOYSA-L
Authentic
808.84
622 to 626nm
Pure
Powder
2353-45-9
85% min.
MFCD00013053
Solubility in water: 150g/L (25°C).
[Na+].[Na+].CCN(CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(C=C1)[C+](C1=CC=C(C=C1)N(CC)CC1=CC=CC(=C1)S([O-])(=O)=O)C1=CC=C(O)C=C1S([O-])(=O)=O
disodium;2-[[4-[ethyl-[(3-sulfonatophenyl)methyl]amino]phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-5-hydroxybenzenesulfonate
16887
808.84
Bordeaux to Maroon
Fast Green FCF
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Suspected of causing genetic defects.

GHS P Statement:
Use personal protective equipment as required.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

SDS
Documents

Documents

RUO – Research Use Only